missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2-Fluorobenzyl)piperazine, 96%

Brand:  Alfa Aesar™ H55888.14

Code : NEW

Additional Details : CAS Number : 89292-78-4 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15432278

  • £298.60 / 25g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

1-(2-Fluorobenzyl)piperazine
89292-78-4
194.25
MFCD02256027

Specifications

1-(2-Fluorobenzyl)piperazine
93° to 96°C (0.2mmHg)
25g
1.5330
96%
MFCD02256027
1.170g/mL
Colorless to pale brown
89292-78-4
Liquid
194.25
Videos
SDS