missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2-Fluorobenzyl)-3-[4-(trifluoromethyl)phenyl]urea, 97%

Brand:  Alfa Aesar™ H61980.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15426906

  • £63.81 / 1g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

1-(2-Fluorobenzyl)-3-[4-(trifluoromethyl)phenyl]urea
312.26
MFCD26143303

Specifications

1-(2-Fluorobenzyl)-3-[4-(trifluoromethyl)phenyl]urea
1g
97%
MFCD26143303
White
Solid
312.26
Videos
SDS