missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Fluorobenzoyl)pyrrolidine, 97%, Thermo Scientific™

Catalog Number 15442808
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
347909-09-5
C11H12FNO
193.221
MFCD00460952
AZBXHPRYPHBYCV-UHFFFAOYSA-N
1-2-fluorobenzoyl pyrrolidine, 2-fluorophenyl 1-pyrrolidinyl methanone, 2-fluorophenyl-pyrrolidin-1-ylmethanone
3497148
(2-fluorophenyl)-pyrrolidin-1-ylmethanone
C1CCN(C1)C(=O)C2=CC=CC=C2F
This item is not returnable. View return policy
347909-09-5
C11H12FNO
193.221
MFCD00460952
AZBXHPRYPHBYCV-UHFFFAOYSA-N
1-2-fluorobenzoyl pyrrolidine, 2-fluorophenyl 1-pyrrolidinyl methanone, 2-fluorophenyl-pyrrolidin-1-ylmethanone
3497148
(2-fluorophenyl)-pyrrolidin-1-ylmethanone
C1CCN(C1)C(=O)C2=CC=CC=C2F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2‘-fluoro-6-pyrrol-2-phenyl-4-quinolone, a derivative of 1-(2-Fluorobenzoyl)pyrrolidine exhibited the most potent cytotoxic activities against renal and melanoma tumor cell lines.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 193.22
Percent Purity 97%
Chemical Name or Material 1-(2-Fluorobenzoyl)pyrrolidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'