missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Fluorobenzoyl)-4-methylpiperidine, 97%, Thermo Scientific™

Catalog Number p-7037729
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
346719-56-0
C13H16FNO
221.275
ZUOUPMRPDVWTSP-UHFFFAOYSA-N
1-2-fluorobenzoyl-4-methylpiperidine, 2-fluorophenyl-4-methylpiperidin-1-yl methanone
3572178
(2-fluorophenyl)-(4-methylpiperidin-1-yl)methanone
CC1CCN(CC1)C(=O)C2=CC=CC=C2F
This item is not returnable. View return policy
346719-56-0
C13H16FNO
221.275
ZUOUPMRPDVWTSP-UHFFFAOYSA-N
1-2-fluorobenzoyl-4-methylpiperidine, 2-fluorophenyl-4-methylpiperidin-1-yl methanone
3572178
(2-fluorophenyl)-(4-methylpiperidin-1-yl)methanone
CC1CCN(CC1)C(=O)C2=CC=CC=C2F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 221.27
Percent Purity 97%
Chemical Name or Material 1-(2-Fluorobenzoyl)-4-methylpiperidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'