missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Fluoro-6-nitrophenyl)-4-piperidone, 97%, Thermo Scientific™

Catalog Number p-7054808
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
2025186-94-9
This item is not returnable. View return policy
2025186-94-9

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 238.22
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(2-Fluoro-6-nitrophenyl)-4-piperidone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'