missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2-Fluoro-6-nitrophenyl)-4-methylpiperazine, 97%

Brand:  Alfa Aesar™ H63175.03

UNSPSC : 12352101

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15419728

  • £52.26 / 1g


Estimated Shipment date
from Supplier 05-10-2017
Add to basket

Specifications

Chemical Identifiers

1-(2-Fluoro-6-nitrophenyl)-4-methylpiperazine
239.25
MFCD13689279

Specifications

1-(2-Fluoro-6-nitrophenyl)-4-methylpiperazine
Solid
239.25
1g
97%
MFCD13689279
Documentation

Technical Documents