missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Fluoro-6-nitrophenyl)-4-methylpiperazine, 97%, Thermo Scientific™

Catalog Number 15458166
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1233952-28-7
C11H14FN3O2
239.25
MFCD13689279
WIAGMIDRHKDTGU-UHFFFAOYSA-N
1-2-fluoro-6-nitrophenyl-4-methylpiperazine
53255582
1-(2-fluoro-6-nitrophenyl)-4-methylpiperazine
CN1CCN(CC1)C2=C(C=CC=C2F)[N+](=O)[O-]
This item is not returnable. View return policy
1233952-28-7
C11H14FN3O2
239.25
MFCD13689279
WIAGMIDRHKDTGU-UHFFFAOYSA-N
1-2-fluoro-6-nitrophenyl-4-methylpiperazine
53255582
1-(2-fluoro-6-nitrophenyl)-4-methylpiperazine
CN1CCN(CC1)C2=C(C=CC=C2F)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 239.25
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(2-Fluoro-6-nitrophenyl)-4-methylpiperazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'