missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2-Fluoro-5-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%

Brand:  Alfa Aesar™ H64194.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15418766

  • £285.14 / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

1-(2-Fluoro-5-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
328.26
MFCD26960695

Specifications

1-(2-Fluoro-5-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
Solid
328.26
5g
97%
MFCD26960695
Videos
SDS