missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2-Fluoro-5-methylphenyl)-3-(4-fluorophenyl)urea, 97%

Brand:  Alfa Aesar™ H62217.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15487166

  • £298.92 / 5g


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

1-(2-Fluoro-5-methylphenyl)-3-(4-fluorophenyl)urea
262.25
MFCD06147104

Specifications

1-(2-Fluoro-5-methylphenyl)-3-(4-fluorophenyl)urea
Solid
262.25
5g
97%
MFCD06147104
Videos
SDS