missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Fluoro-4-nitrophenyl)pyrrolidine, 97%, Thermo Scientific™

Catalog Number p-7055358
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
385380-74-5
C10H11FN2O2
210.208
SMKXLBISYVJESG-UHFFFAOYSA-N
1-2-fluoro-4-nitrophenyl pyrrolidine, 3-fluoro-4-pyrrolidin-1-yl nitrobenzene, pyrrolidine, 1-2-fluoro-4-nitrophenyl, pubchem18700, cambridge id 5842403, 2-fluoro-4-nitrophenyl pyrrolidine, 1-2-fluoro-4-nitro-phenyl pyrrolidine
2872122
1-(2-fluoro-4-nitrophenyl)pyrrolidine
C1CCN(C1)C2=C(C=C(C=C2)[N+](=O)[O-])F
This item is not returnable. View return policy
385380-74-5
C10H11FN2O2
210.208
SMKXLBISYVJESG-UHFFFAOYSA-N
1-2-fluoro-4-nitrophenyl pyrrolidine, 3-fluoro-4-pyrrolidin-1-yl nitrobenzene, pyrrolidine, 1-2-fluoro-4-nitrophenyl, pubchem18700, cambridge id 5842403, 2-fluoro-4-nitrophenyl pyrrolidine, 1-2-fluoro-4-nitro-phenyl pyrrolidine
2872122
1-(2-fluoro-4-nitrophenyl)pyrrolidine
C1CCN(C1)C2=C(C=C(C=C2)[N+](=O)[O-])F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 210.2
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(2-Fluoro-4-nitrophenyl)pyrrolidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'