missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2-Fluoro-4-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%

Brand:  Alfa Aesar™ H64251.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15418806

  • £51.94 / 1g


Estimated Shipment date
from Supplier 03-05-2018
Add to basket

Specifications

Chemical Identifiers

1-(2-Fluoro-4-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
328.26
MFCD26960696

Specifications

1-(2-Fluoro-4-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
Solid
328.26
1g
97%
MFCD26960696
Videos
SDS