missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2-Fluoro-3-methylphenyl)-3-(4-fluorophenyl)urea, 97%

Brand:  Alfa Aesar™ H62467.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15447466

  • £53.11 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

1-(2-Fluoro-3-methylphenyl)-3-(4-fluorophenyl)urea
262.25
MFCD26143327

Specifications

1-(2-Fluoro-3-methylphenyl)-3-(4-fluorophenyl)urea
Solid
262.25
1g
97%
MFCD26143327
Videos
SDS