missing translation for 'onlineSavingsMsg'
Learn More

1-(2'-Fluoro[1,1'-biphenyl]-4-yl)propan-1-one, 97%, Thermo Scientific™

Catalog Number p-2706173
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
37989-92-7
C15H13FO
228.266
MFCD00236612
ZXIQZWXVSBJPTC-UHFFFAOYSA-N
1-2'-fluoro-1,1'-biphenyl-4-yl propan-1-one, 1-2'-fluoro 1,1'-biphenyl-4-yl propan-1-one, 1-4-2-fluorophenyl phenyl propan-1-one, 4'-2-fluorophenyl propiophenone, 4-2-fluorophenyl propiophenone, 1-propanone,1-2'-fluoro 1,1'-biphenyl-4-yl, maybridge3_006207, 2-fluoro-4'-propanoylbiphenyl
2737490
1-[4-(2-fluorophenyl)phenyl]propan-1-one
CCC(=O)C1=CC=C(C=C1)C2=CC=CC=C2F
This item is not returnable. View return policy
37989-92-7
C15H13FO
228.266
MFCD00236612
ZXIQZWXVSBJPTC-UHFFFAOYSA-N
1-2'-fluoro-1,1'-biphenyl-4-yl propan-1-one, 1-2'-fluoro 1,1'-biphenyl-4-yl propan-1-one, 1-4-2-fluorophenyl phenyl propan-1-one, 4'-2-fluorophenyl propiophenone, 4-2-fluorophenyl propiophenone, 1-propanone,1-2'-fluoro 1,1'-biphenyl-4-yl, maybridge3_006207, 2-fluoro-4'-propanoylbiphenyl
2737490
1-[4-(2-fluorophenyl)phenyl]propan-1-one
CCC(=O)C1=CC=C(C=C1)C2=CC=CC=C2F
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula FC6H4C6H4C(O)CH2CH3
Quantity 1 g
Formula Weight 228.26
Percent Purity 97%
Chemical Name or Material 1-(2'-Fluoro[1, 1'-biphenyl]-4-yl)propan-1-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'