missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxypropane, Extra Pure, SLR, Fisher Chemical™

Brand:  Fisher Chemical E/0050/PB17

Code : 21

Additional Details : CAS Number : 75-56-9 Weight : 2.50000kg Transport : UN number : 1280 Chem class : 3 Pack group : I


Quantity 2.5L

 View more versions of this product

Product Code. 10060140

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Chemical Identifiers

75-56-9
58.08
GOOHAUXETOMSMM-UHFFFAOYNA-N
6378
2-methyloxirane
C3H6O
MFCD00005126
propylene oxide, 1,2-epoxypropane, methyloxirane, epoxypropane, propyleneoxide, propene oxide, oxirane, methyl, 1,2-propylene oxide, propylene epoxide, 2,3-epoxypropane
CHEBI:38685
CC1CO1

Specifications

75-56-9
-112°C
C3H6O
Plastic Coated Glass bottle
2.5L
CC1CO1
58.08
CHEBI:38685
Liquid
590mbar at 20°C
Colorless
34°C
MFCD00005126
propylene oxide, 1,2-epoxypropane, methyloxirane, epoxypropane, propyleneoxide, propene oxide, oxirane, methyl, 1,2-propylene oxide, propylene epoxide, 2,3-epoxypropane
GOOHAUXETOMSMM-UHFFFAOYNA-N
2-methyloxirane
6378
58.08g/mol
2
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1,2-Epoxypropane

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 1
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 1B
  • Carcinogenicity Category 1B
  • Specific target organ toxicity Category 3

Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H340-May cause genetic defects.
  • H350-May cause cancer.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

SDS Documents
Certificates
Special Offers

Special Offers