missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxy-3-methylbutane, 98+%, Thermo Scientific Chemicals

Catalog Number p-7047470
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1438-14-8
C5H10O
86.134
MFCD02683501
REYZXWIIUPKFTI-UHFFFAOYSA-N
2-isopropyloxirane, 1,2-epoxy-3-methylbutane, isopropyloxirane, oxirane, 1-methylethyl, butane, 1,2-epoxy-3-methyl, 2-propan-2-yl oxirane, oxirane,2-1-methylethyl, 1-methylethyl-oxirane, isopropylethylene oxide, acmc-1asns
102618
2-propan-2-yloxirane
CC(C)C1CO1
This item is not returnable. View return policy
1438-14-8
C5H10O
86.134
MFCD02683501
REYZXWIIUPKFTI-UHFFFAOYSA-N
2-isopropyloxirane, 1,2-epoxy-3-methylbutane, isopropyloxirane, oxirane, 1-methylethyl, butane, 1,2-epoxy-3-methyl, 2-propan-2-yl oxirane, oxirane,2-1-methylethyl, 1-methylethyl-oxirane, isopropylethylene oxide, acmc-1asns
102618
2-propan-2-yloxirane
CC(C)C1CO1

1,2-Epoxy-3-methylbutane is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Epoxy-3-methylbutane is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store in cool place. Store under inert gas. It is sensitive to moisture. Incompatible with bases and oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 0.815
Boiling Point 72°C to 75°C
Flash Point −12°C (10°F)
Odor Recognizable
Refractive Index 1.3915
Quantity 1 g
UN Number UN1993
Beilstein 102418
Sensitivity Moisture sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 86.13
Percent Purity ≥98%
Chemical Name or Material 1,2-Epoxy-3-methylbutane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'