missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxy-3,3,3-trifluoropropane, 98%, Thermo Scientific Chemicals

Catalog Number p-7045222
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
359-41-1
C3H3F3O
112.051
MFCD00041506
AQZRARFZZMGLHL-UHFFFAOYSA-N
1,1,1-trifluoro-2,3-epoxypropane, 2-trifluoromethyl oxirane, 3,3,3-trifluoro-1,2-epoxypropane, 1,2-epoxy-3,3,3-trifluoropropane, oxirane, trifluoromethyl, 3,3,3-trifluoropropane epoxide, pubchem20840, trifluoromethyl oxirane, acmc-20apk3
520769
2-(trifluoromethyl)oxirane
C1C(O1)C(F)(F)F
This item is not returnable. View return policy
359-41-1
C3H3F3O
112.051
MFCD00041506
AQZRARFZZMGLHL-UHFFFAOYSA-N
1,1,1-trifluoro-2,3-epoxypropane, 2-trifluoromethyl oxirane, 3,3,3-trifluoro-1,2-epoxypropane, 1,2-epoxy-3,3,3-trifluoropropane, oxirane, trifluoromethyl, 3,3,3-trifluoropropane epoxide, pubchem20840, trifluoromethyl oxirane, acmc-20apk3
520769
2-(trifluoromethyl)oxirane
C1C(O1)C(F)(F)F

1,2-Epoxy-3,3,3-trifluoropropane is used for the synthesis of trifluorolactic acid. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Epoxy-3,3,3-trifluoropropane is used for the synthesis of trifluorolactic acid. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Not miscible with water.

Notes
Moisture Sensitive. Protect from humidity and water. Incompatible materials are heat, oxidizing agents and water. Store away from moisture.
TRUSTED_SUSTAINABILITY
Density 1.3
Boiling Point 38°C to 40°C
Flash Point −40°C (−40°F)
Refractive Index 1.297
Quantity 1 g
UN Number UN1992
Beilstein 103730
Sensitivity Moisture sensitive
Solubility Information Not miscible with water.
Formula Weight 112.05
Percent Purity 98%
Chemical Name or Material 1,2-Epoxy-3,3,3-trifluoropropane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'