Learn More
1,2-Dipiperidinoethane, 98%, ACROS Organics™
Brand: Acros Organics 185110050
Code : 29
Additional Details : CAS Number : 1932-04-3 Weight : 0.05500kg
Quantity | 5g |
---|
Chemical Identifiers
1, 2-Dipiperidinoethane | |
97.5 | |
≥97.5% (GC) | |
C12H24N2 | |
MFCD00014644 |
98% | |
100.0 | |
98% | |
196.33 | |
1, 1'-(1, 2-Ethanediyl)bispiperidine |
Specifications
1, 2-Dipiperidinoethane | |
Clear | |
98% | |
100.0 | |
1.4865 to 1.4885 (20°C, 589nm) | |
≥97.5% (GC) | |
C12H24N2 | |
MFCD00014644 | |
1, 1'-(1, 2-Ethanediyl)bispiperidine |
0.91g/mL | |
5g | |
97.5 | |
Authentic | |
Liquid | |
98% | |
196.33 | |
20, I, 19 |