missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Diiodoethane, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
1,2-Diiodoethane is used as C-X bond formation reagent. It is also used as Pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,2-Diiodoethane is used as C-X bond formation reagent. It is also used as Pharmaceutical intermediates.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Store at 4°C. Air & Light Sensitive. Incompatible with strong bases, strong oxidizing agents, heat and light. Protect from heat.
1,2-Diiodoethane is used as C-X bond formation reagent. It is also used as Pharmaceutical intermediates.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Store at 4°C. Air & Light Sensitive. Incompatible with strong bases, strong oxidizing agents, heat and light. Protect from heat.
Specifications
Melting Point | 80°C to 84°C |
Density | 2.13 |
Quantity | 5 g |
Beilstein | 1730870 |
Sensitivity | Light sensitive |
Solubility Information | Soluble in methanol. Insoluble in water. |
Formula Weight | 281.86 |
Percent Purity | 98% |
Chemical Name or Material | 1,2-Diiodoethane |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'