missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diiodobenzene, 98%, Thermo Scientific Chemicals

Catalog Number 10656032
Click to view available options
Quantity:
5 g
20 g
Packaging:
Glass bottle
This item is not returnable. View return policy
615-42-9
C6H4I2
329.91
MFCD00001038
BBOLNFYSRZVALD-UHFFFAOYSA-N
o-diiodobenzene, benzene, 1,2-diiodo, benzene, o-diiodo, diiodobenzene, benzene, diiodo, benzene,2-diiodo, pubchem3831, 1,2-diiodobenzene
11994
1,2-diiodobenzene
IC1=CC=CC=C1I
This item is not returnable. View return policy
615-42-9
C6H4I2
329.91
MFCD00001038
BBOLNFYSRZVALD-UHFFFAOYSA-N
o-diiodobenzene, benzene, 1,2-diiodo, benzene, o-diiodo, diiodobenzene, benzene, diiodo, benzene,2-diiodo, pubchem3831, 1,2-diiodobenzene
11994
1,2-diiodobenzene
IC1=CC=CC=C1I

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Brown to Yellow
Density 2.5200g/mL
Boiling Point 152°C (15.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H4I2
Refractive Index 1.7176
Quantity 5 g
Beilstein 05, 225
Specific Gravity 2.52
Formula Weight 329.9
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 2-Diiodobenzene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,2-Diiodobenzene
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'