missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dicyanobenzene, 98%, Thermo Scientific Chemicals

Catalog Number p-107127
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
91-15-6
C8H4N2
128.13
MFCD00001771
XQZYPMVTSDWCCE-UHFFFAOYSA-N
phthalonitrile, 1,2-dicyanobenzene, o-phthalodinitrile, phthalodinitrile, 1,2-benzenedicarbonitrile, o-dicyanobenzene, 1,2-benzodinitrile, o-pdn, o-cyanobenzonitrile, o-benzenedicarbonitrile
7042
benzene-1,2-dicarbonitrile
N#CC1=CC=CC=C1C#N
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
91-15-6
C8H4N2
128.13
MFCD00001771
XQZYPMVTSDWCCE-UHFFFAOYSA-N
phthalonitrile, 1,2-dicyanobenzene, o-phthalodinitrile, phthalodinitrile, 1,2-benzenedicarbonitrile, o-dicyanobenzene, 1,2-benzodinitrile, o-pdn, o-cyanobenzonitrile, o-benzenedicarbonitrile
7042
benzene-1,2-dicarbonitrile
N#CC1=CC=CC=C1C#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 138.0°C to 142.0°C
Color White to Beige
Density 1.2400g/mL
Flash Point >130°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula C6H4(CN)2
Quantity 250 g
Beilstein 09, 815
Specific Gravity 1.24
Solubility Information Solubility in water: 0.56g/L (25°C). Other solubilities: soluble in acetone and benzene,soluble in ether and alcohol
Formula Weight 128.13
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 1, 2-Dicyanobenzene, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'