missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloropropane, 98%, pure, Thermo Scientific Chemicals

Catalog Number p-105830
Click to view available options
Quantity:
5 mL
50 mL
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
78-87-5
C3H6Cl2
112.98
MFCD00000868
KNKRKFALVUDBJE-UHFFFAOYNA-N
propylene dichloride, propane, 1,2-dichloro, propylene chloride, dwuchloropropan, bichlorure de propylene, rcra waste number u083, caswell no. 324, dwuchloropropan polish, alpha,beta-dichloropropane, ccris 951
6564
CHEBI:82163
1,2-dichloropropane
CC(Cl)CCl
This item is not returnable. View return policy
78-87-5
C3H6Cl2
112.98
MFCD00000868
KNKRKFALVUDBJE-UHFFFAOYNA-N
propylene dichloride, propane, 1,2-dichloro, propylene chloride, dwuchloropropan, bichlorure de propylene, rcra waste number u083, caswell no. 324, dwuchloropropan polish, alpha,beta-dichloropropane, ccris 951
6564
CHEBI:82163
1,2-dichloropropane
CC(Cl)CCl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point -100.0°C
Color Colorless to Yellow
Density 1.1560g/mL
Boiling Point 95.0°C to 96.0°C
Flash Point 15°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(Cl)CH2Cl
Refractive Index 1.4380 to 1.4400
Quantity 5 mL
Beilstein 01, 105
Merck Index 15, 7967
Specific Gravity 1.156
Solubility Information Solubility in water: 3g/L (20°C). Other solubilities: soluble in most common organic solvents
Formula Weight 112.99
Percent Purity 98%
Grade Pure
Physical Form Liquid
Chemical Name or Material 1, 2-Dichloropropane, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'