missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dichloroethane, Spectrophotometric Grade, 99+%, Thermo Scientific Chemicals
38.40 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
Description
1,2-dichloroethane is used in the production of vinyl chloride which is used to make a variety of plastic and vinyl products including polyvinyl chloride (PVC) pipes, furniture and automobile upholstery, wall coverings, house wares, and automobile parts.1,2-Dichloroethane is also used generally as an intermediate for other organic chemical compounds and as a solvent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -35°C |
Density | 1.256 g/mL |
Boiling Point | 83°C |
Quantity | 1 L |
Flash Point | 15°C (59°F) |
Physical Form | Liquid |
Refractive Index | 1.4448 |
UN Number | UN1184 |
Beilstein | 605264 |
Merck Index | 14,3797 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'