missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloroethane, ACS, 99+%, Thermo Scientific Chemicals

Catalog Number 15431517
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 1 L
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
107-06-2
C2H4Cl2
98.95
MFCD00000963
WSLDOOZREJYCGB-UHFFFAOYSA-N
ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion
11
CHEBI:27789
1,2-dichloroethane
ClCCCl
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
107-06-2
C2H4Cl2
98.95
MFCD00000963
WSLDOOZREJYCGB-UHFFFAOYSA-N
ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion
11
CHEBI:27789
1,2-dichloroethane
ClCCCl

Maximum level of impurities: Color (APHA) 10, Evaporation residue 0.002%, Titratable acid 0.0003meq/g, H2O 0.03%

Used to make vinyl chloride, and chlorinated solvents(trichloroethane, trichloroethylene, perchloroethylene, and vinylidene), remove grease, resins, glue and dirt. Used as a solvent in the manufacturing of polystyrene and SBR latex. It is added to leaded petrol as an anti-knock compound. It is also used in polyvinyl chloride (PVC) pipes, packaging materials, furniture, auto mobile parts, wall coverings and housewares. It is also used generally as an intermediate for other organic chemical compounds and as a solvent. Used as fumigant residues for wheat, flour, bran, middlings and bread. Used as solvent for the isomer distribution in the acetylation of benzene and toluene under the influence of aluminum chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -35°C
Density 1.256
Boiling Point 83°C
Quantity 4 x 1 L
Flash Point 15°C (59°F)
Physical Form Liquid
Refractive Index 1.4448
UN Number UN1184
Beilstein 605264
Merck Index 14,3797
Solubility Information Miscible in methanol,diethyl ether,n-octanol,acetone. Very slightly miscible in water.
Formula Weight 98.96
Percent Purity ≥99%
Grade ACS
Chemical Name or Material 1,2-Dichloroethane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'