missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dichloroethane, 99.8+%, extra pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
1 L
2.5 L
25 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Orange to Yellow |
Density | 1.2500g/mL |
Quantity | 2.5 L |
Infrared Spectrum | Authentic |
Packaging | Glass Bottle |
Physical Form | Crystalline Powder |
Refractive Index | 1.4440 to 1.4460 (20°C, 589nm) |
Assay Percent Range | 99.8+% |
Residue after Evaporation | 0.005% max. |
Linear Formula | ClCH2CH2Cl |
Show More |
Safety and Handling
Product Identifier
- 1,2-Dichloroethane
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 4
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Carcinogenicity Category 1B
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H350-May cause cancer.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P311-Call a POISON CENTER/doctor/ .
- P362-Take off contaminated clothing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'