missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dichlorobenzene, 99%, Reagent Grade, Honeywell™
Click to view available options
Quantity:
100 mL
1 L
2.5 L
20 L
Packaging:
Glass bottle
Stainless steel can
Description
- ≥95% for general laboratory use.
Specifications
Melting Point | -17°C |
Color | Colorless to Yellow |
Density | 1.306g/mL at 25°C |
Boiling Point | 180°C (1013 hPa) |
Explosion Limit | 0.092 |
Flash Point | 66°C (150.8°F) |
Ignition Point | 1198°F |
Odor | Aromatic |
Packaging | Glass bottle |
Quantity | 2.5 L |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'