missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloro-4-fluoro-5-nitrobenzene, 95%, Thermo Scientific Chemicals

Catalog Number 15402139
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
2339-78-8
C6H2Cl2FNO2
209.985
MFCD00075330
FXOCDIKCKFOUDE-UHFFFAOYSA-N
4,5-dichloro-2-fluoronitrobenzene, benzene, 1,2-dichloro-4-fluoro-5-nitro, buttpark 89\07-58, 4,5-dichloro-2-fluoro-1-nitrobenzene, pubchem4385, intermediates-zcf02626, ksc201k5p, 1,2-ichloro-4-luoro-5-itro-benzene
533951
1,2-dichloro-4-fluoro-5-nitrobenzene
C1=C(C(=CC(=C1Cl)Cl)F)[N+](=O)[O-]
This item is not returnable. View return policy
2339-78-8
C6H2Cl2FNO2
209.985
MFCD00075330
FXOCDIKCKFOUDE-UHFFFAOYSA-N
4,5-dichloro-2-fluoronitrobenzene, benzene, 1,2-dichloro-4-fluoro-5-nitro, buttpark 89\07-58, 4,5-dichloro-2-fluoro-1-nitrobenzene, pubchem4385, intermediates-zcf02626, ksc201k5p, 1,2-ichloro-4-luoro-5-itro-benzene
533951
1,2-dichloro-4-fluoro-5-nitrobenzene
C1=C(C(=CC(=C1Cl)Cl)F)[N+](=O)[O-]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 17°C
Color Yellow
Density 1.596 g/mL
Boiling Point 247°C
Flash Point >113°C (235°F)
Refractive Index 1.575
Quantity 5 g
Beilstein 3269124
Formula Weight 209.99
Percent Purity 95%
Physical Form Low Melting Solid
Chemical Name or Material 1,2-Dichloro-4-fluoro-5-nitrobenzene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'