missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloro-3-iodobenzene, 97%, ACROS Organics™

Brand:  Acros Organics 222011000

Code : 29

Additional Details : CAS Number : 2401-21-0 Weight : 0.15000kg


Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10732685

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

1, 2-Dichloro-3-iodobenzene
2401-21-0
100.0
97%
Cl2C6H3I
MFCD00001034
97%
96.0
97%
C6H3Cl2I
272.9

Specifications

1, 2-Dichloro-3-iodobenzene
123.0° to 126.0°C (20.0 mmHg)
33.0° to 37.0°C
97%
96.0
Authentic
97%
C6H3Cl2I
272.9
Glass bottle
>110°C
White
100g
2401-21-0
100.0
Crystalline Mass
97%
Cl2C6H3I
MFCD00001034
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1,2-Dichloro-3-iodobenzene

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS