missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromobenzene, 98%, Thermo Scientific Chemicals

Catalog Number p-3736123
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
583-53-9
C6H4Br2
235.91
MFCD00000057
WQONPSCCEXUXTQ-UHFFFAOYSA-N
o-dibromobenzene, dibromobenzene, benzene, 1,2-dibromo, ortho-dibromobenzene, benzene, o-dibromo, benzene, dibromo, unii-52k9w7u6eh, dibromobenzene mixed isomers, 1,2 dibromobenzene, 1,6-dibromobenzene
11414
CHEBI:37152
1,2-dibromobenzene
C1=CC=C(C(=C1)Br)Br
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
583-53-9
C6H4Br2
235.91
MFCD00000057
WQONPSCCEXUXTQ-UHFFFAOYSA-N
o-dibromobenzene, dibromobenzene, benzene, 1,2-dibromo, ortho-dibromobenzene, benzene, o-dibromo, benzene, dibromo, unii-52k9w7u6eh, dibromobenzene mixed isomers, 1,2 dibromobenzene, 1,6-dibromobenzene
11414
CHEBI:37152
1,2-dibromobenzene
C1=CC=C(C(=C1)Br)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 4.0°C to 6.0°C
Color Colorless to Yellow or Pink
Density 1.9560g/mL
Boiling Point 224.0°C
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H4Br2
Refractive Index 1.6100 to 1.6120
Quantity 25 mL
Beilstein 05, 210
Specific Gravity 1.956
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol,acetone,ether,benzene,ca,rbon tetrachloride,n-heptane
Formula Weight 235.91
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 2-Dibromobenzene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,2-Dibromobenzene
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'