1,2-Dibromo-1,1,2-trichloroethane, 99%, ACROS Organics™


Overview
£6.72 valid until 31/03/2017
Log in to get the promotional price



Brand: Acros Organics

Manufacturer Part Number: 279030250

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 13749-38-7 Weight: 0.07500kg


Packaging Glass bottle
Quantity 25g

Product Code. 10477382

Quantity Price
1 £ 16.8 / 25g
EU Stock 2
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 1,2-Dibromo-1,1,2-trichloroethane
Name Note 99%
CAS 13749-38-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
MDL Number MFCD00018841

Specification

Physical Form Liquid
Assay ≥98.5%
Quantity 25g
Color Clear colorless
CAS Min % 98.5
Infrared Spectrum Authentic
CAS 13749-38-7
Assay Percent Range 99%
Refractive Index 1.5700 to 1.5720 (20°C, 589nm)
MDL Number MFCD00018841
Packaging Glass bottle
CAS Max % 100.0
Name Note 99%
Chemical Name or Material 1,2-Dibromo-1,1,2-trichloroethane
Documentation

Technical documents

Special Offers