missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromo-1,1,2-trichloroethane, 99%, Thermo Scientific™

Catalog Number p-2689443
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
13749-38-7
C2HBr2Cl3
291.2
MFCD00018841
VXKOVUFOVCJHBA-UHFFFAOYSA-N
ethane,1,2-dibromo-1,1,2-trichloro, acmc-20ap6p, 1,2-dibromotrichloro ethane
114562
1,2-dibromo-1,1,2-trichloroethane
C(C(Cl)(Cl)Br)(Cl)Br
This item is not returnable. View return policy
13749-38-7
C2HBr2Cl3
291.2
MFCD00018841
VXKOVUFOVCJHBA-UHFFFAOYSA-N
ethane,1,2-dibromo-1,1,2-trichloro, acmc-20ap6p, 1,2-dibromotrichloro ethane
114562
1,2-dibromo-1,1,2-trichloroethane
C(C(Cl)(Cl)Br)(Cl)Br
CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Flash Point >200°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5700 to 1.5720
Quantity 5 g
Formula Weight 291.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1,2-Dibromo-1,1,2-trichloroethane, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'