missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diamino-4,5-difluorobenzene, 97%, Thermo Scientific Chemicals

Catalog Number 15284774
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
76179-40-3
C6H6F2N2
144.13
MFCD00061131
PPWRHKISAQTCCG-UHFFFAOYSA-N
1,2-diamino-4,5-difluorobenzene, 4,5-difluoro-1,2-phenylenediamine, 4,5-difluoro-o-phenylenediamine, 2-amino-4,5-difluorophenylamine, 4,5-difluoro-benzene-1,2-diamine, 4,5-difluoro-1,2-benzenediamine, 1,2-benzenediamine,4,5-difluoro, 1,2-benzenediamine, 4,5-difluoro, pubchem4354, 2-amino-4,5-difluoroaniline
2736755
4,5-difluorobenzene-1,2-diamine
NC1=CC(F)=C(F)C=C1N
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
76179-40-3
C6H6F2N2
144.13
MFCD00061131
PPWRHKISAQTCCG-UHFFFAOYSA-N
1,2-diamino-4,5-difluorobenzene, 4,5-difluoro-1,2-phenylenediamine, 4,5-difluoro-o-phenylenediamine, 2-amino-4,5-difluorophenylamine, 4,5-difluoro-benzene-1,2-diamine, 4,5-difluoro-1,2-benzenediamine, 1,2-benzenediamine,4,5-difluoro, 1,2-benzenediamine, 4,5-difluoro, pubchem4354, 2-amino-4,5-difluoroaniline
2736755
4,5-difluorobenzene-1,2-diamine
NC1=CC(F)=C(F)C=C1N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 130°C to 133°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (F)C6H2(NH2)2
Quantity 5 g
Formula Weight 144.12
Percent Purity 97%
Chemical Name or Material 1, 2-Diamino-4, 5-difluorobenzene
compliance-icons
Product Identifier
  • 1,2-Diamino-4,5-difluorobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'