missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2-Cyanoethyl)indole-6-carboxylic acid, 97%

Brand:  Alfa Aesar™ H63216.03

Code : NEW

Additional Details : CAS Number : 885266-69-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15449788

  • £95.61 / 1g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

1-(2-Cyanoethyl)indole-6-carboxylic acid
885266-69-3
214.22
MFCD04973976

Specifications

1-(2-Cyanoethyl)indole-6-carboxylic acid
885266-69-3
97%
MFCD04973976
1g
Solid
214.22
Videos
SDS