missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2-Cyanoethyl)indole-6-carboxylic acid, 97%

Brand:  Alfa Aesar™ H63216.06

Code : NEW

Additional Details : CAS Number : 885266-69-3 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15459788

  • £478.06 / 5g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

1-(2-Cyanoethyl)indole-6-carboxylic acid
885266-69-3
214.22
MFCD04973976

Specifications

1-(2-Cyanoethyl)indole-6-carboxylic acid
885266-69-3
97%
MFCD04973976
5g
Solid
214.22
Videos
SDS