missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2-Chlorophenyl)-3-(4-fluorophenyl)urea, 97%

Brand:  Alfa Aesar™ H62129.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15418708

  • £398.56 / 5g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

1-(2-Chlorophenyl)-3-(4-fluorophenyl)urea
264.68
MFCD00018186

Specifications

1-(2-Chlorophenyl)-3-(4-fluorophenyl)urea
Solid
264.68
5g
97%
MFCD00018186
Videos
SDS