missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2-Chlorophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea, 97%

Brand:  Alfa Aesar™ H65119.03

Code : NEW

Additional Details : CAS Number : 418805-03-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15449386

  • £85.44 / 1g


Estimated Shipment date
from Supplier 11-05-2018
Add to basket

Specifications

Chemical Identifiers

1-(2-Chlorophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea
349.13
1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(2-chlorophenyl)urea
418805-03-5
MFCD01654414

Specifications

1-(2-Chlorophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea
418805-03-5
97%
MFCD01654414
1g
Solid
349.13
1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(2-chlorophenyl)urea
Videos
SDS