missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2-Chloro-6-nitrophenyl)azepane, 97%

Brand:  Alfa Aesar™ H63847.03

Code : NEW

Additional Details : Weight : 0.00100kg Transport : UN number : 3259 Chem class : 1b Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15440769

  • £50.70 / 1g


Estimated Shipment date
from Supplier 24-05-2018
Add to basket

Specifications

Chemical Identifiers

1-(2-Chloro-6-nitrophenyl)azepane
254.71
UN3259
1-(2-Chloro-6-nitrophenyl)hexamethyleneimine

Specifications

1-(2-Chloro-6-nitrophenyl)azepane
Solid
254.71
1-(2-Chloro-6-nitrophenyl)hexamethyleneimine
1g
97%
UN3259
Videos
SDS