missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2-Chloro-6-nitrophenyl)-4-piperidone, 97%

Brand:  Alfa Aesar™ H63085.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15419578

  • £101.50 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

1-(2-Chloro-6-nitrophenyl)-4-piperidone
254.67

Specifications

1-(2-Chloro-6-nitrophenyl)-4-piperidone
1925644-19-4
97%
1g
Solid
254.67
Videos
SDS