missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2-Chloro-4-nitrophenyl)pyrrolidine, 97%

Brand:  Alfa Aesar™ H63415.03

Code : NEW

Additional Details : CAS Number : 55403-91-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15420099

  • £50.70 / 1g


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

1-(2-Chloro-4-nitrophenyl)pyrrolidine
55403-91-3
226.66

Specifications

1-(2-Chloro-4-nitrophenyl)pyrrolidine
55403-91-3
97%
1g
Solid
226.66
Videos
SDS