missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 1-(2-Chloro-4-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%
Brand: Alfa Aesar™ H64882.03
Code : NEW
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
1-(2-Chloro-4-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea | |
344.72 | |
MFCD26960713 |
Specifications
1-(2-Chloro-4-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea | |
97% | |
344.72 |
Solid | |
1g | |
MFCD26960713 |