missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2-Chloro-4-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%

Brand:  Alfa Aesar™ H64882.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15469226

  • £351.92 / 5g


Estimated Shipment date
from Supplier 06-07-2018
Add to basket

Specifications

Chemical Identifiers

1-(2-Chloro-4-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
344.72
MFCD26960713

Specifications

1-(2-Chloro-4-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
97%
344.72
Solid
5g
MFCD26960713
Videos
SDS