missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 1-(2-Chloro-4-methylphenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea, 97%
Brand: Alfa Aesar™ H65241.03
Code : NEW
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
1-(2-Chloro-4-methylphenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea | |
363.16 | |
MFCD27975741 |
Specifications
1-(2-Chloro-4-methylphenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea | |
97% | |
363.16 |
Solid | |
1g | |
MFCD27975741 |