missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2-Carbamoylethyl)indole-6-carboxylic acid, 97%

Brand:  Alfa Aesar™ H63934.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15410919

  • £155.82 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

1-(2-Carbamoylethyl)indole-6-carboxylic acid
232.24
1-(3-Amino-3-oxopropyl)-indole-6-carboxylic acid
885266-81-9
MFCD06657683

Specifications

1-(2-Carbamoylethyl)indole-6-carboxylic acid
885266-81-9
97%
MFCD06657683
1g
Solid
232.24
1-(3-Amino-3-oxopropyl)-indole-6-carboxylic acid
Videos
SDS