missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2-Bromo-4-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%

Brand:  Alfa Aesar™ H64325.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15428846

  • £44.31 / 1g


Estimated Shipment date
from Supplier 30-01-2018
Add to basket

Specifications

Chemical Identifiers

1-(2-Bromo-4-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
389.17
MFCD26960698

Specifications

1-(2-Bromo-4-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
1g
389.17
Solid
97%
MFCD26960698
Videos
SDS