missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2-Bromo-4-methylphenyl)-3-(4-fluorophenyl)urea, 97%

Brand:  Alfa Aesar™ H62192.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15467146

  • £239.56 / 5g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

1-(2-Bromo-4-methylphenyl)-3-(4-fluorophenyl)urea
323.16
MFCD07096409

Specifications

1-(2-Bromo-4-methylphenyl)-3-(4-fluorophenyl)urea
5g
323.16
Solid
97%
MFCD07096409
Videos
SDS