missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(methanesulfonamido)benzene, 97%, Thermo Scientific Chemicals

Catalog Number p-7024694
Click to view available options
Quantity:
2.5 g
10 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7596-80-7
C8H12N2O4S2
264.31
MFCD00276605
BYZJRLRCNOECEV-UHFFFAOYSA-N
1,2-bis methanesulfonamido benzene, n,n'-1,2-phenylene dimethanesulfonamide, n-2-methanesulfonamidophenyl methanesulfonamide, n-2-methylsulfonyl amino phenyl methanesulfonamide, methylsulfonyl 2-methylsulfonyl amino phenyl amine, maybridge1_000065, n,n'-o-phenylenebis methanesulfonamide, o-phenylenediamine n,n'-bis methanesulfonamide, n-2-methanesulfonamido phenyl methanesulfonamide
227307
N-[2-(methanesulfonamido)phenyl]methanesulfonamide
CS(=O)(=O)NC1=CC=CC=C1NS(C)(=O)=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7596-80-7
C8H12N2O4S2
264.31
MFCD00276605
BYZJRLRCNOECEV-UHFFFAOYSA-N
1,2-bis methanesulfonamido benzene, n,n'-1,2-phenylene dimethanesulfonamide, n-2-methanesulfonamidophenyl methanesulfonamide, n-2-methylsulfonyl amino phenyl methanesulfonamide, methylsulfonyl 2-methylsulfonyl amino phenyl amine, maybridge1_000065, n,n'-o-phenylenebis methanesulfonamide, o-phenylenediamine n,n'-bis methanesulfonamide, n-2-methanesulfonamido phenyl methanesulfonamide
227307
N-[2-(methanesulfonamido)phenyl]methanesulfonamide
CS(=O)(=O)NC1=CC=CC=C1NS(C)(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 211°C to 214°C
Odor Odorless
Quantity 2.5 g
Formula Weight 264.32
Percent Purity 97%
Chemical Name or Material 1,2-Bis(methanesulfonamido)benzene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'