missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(dipentafluorophenylphosphino)ethane, 95%, Acros Organics™

Brand:  Acros Organics 363700010

Code : 29

Additional Details : CAS Number : 76858-94-1 Weight : 0.05100kg


Quantity 1g

 View more versions of this product

Product Code. 10394112

  • £77.90 / 1g


Estimated Shipment
14-02-2018
Add to basket


Specifications

Chemical Identifiers

1, 2-Bis(dipentafluorophenylphosphino)ethane
95%
758.26
95%
C26H4F20P2

Specifications

1, 2-Bis(dipentafluorophenylphosphino)ethane
Authentic
Crystalline Powder
95%
758.26
190.0° to 195.0°C
76858-94-1
Glass bottle
95%
C26H4F20P2
White to light beige
1g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1,2-Bis(dipentafluorophenylphosphino)ethane

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS