missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,2-Bis(dimethylsilyl)benzene, 98%

Brand:  Alfa Aesar™ 043755.09

Code : NEW

Additional Details : CAS Number : 17985-72-7 Weight : 0.01000kg

Quantity 10g

 View more versions of this product

Product Code. 11320378

  • £367.60 / 10g


Estimated Shipment date
from Supplier 30-01-2018
Add to basket

Specifications

Chemical Identifiers

1,2-Bis (dimethylsilyl)benzene
C10H18Si2
MFCD00142462
17985-72-7
194.43

Specifications

1,2-Bis (dimethylsilyl)benzene
Liquid
1.5140
194.43
0.898g/mL
10g
17985-72-7
98%
C10H18Si2
MFCD00142462
101°C (13mmHg)
70°C (158°F)
Videos
SDS