missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Bis(diethylphosphino)ethane, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
It is used in chemical reaearch, organic Intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Specifications
Color | Colorless |
Density | 0.884 g/mL |
Boiling Point | 124°C to 126°C (10mmHg) |
Odor | Pungent |
Linear Formula | (CH3CH2)2PCH2CH2P(CH2CH3)2 |
Quantity | 1 g |
UN Number | UN2845 |
Sensitivity | Air Sensitive |
Solubility Information | It is soluble in organic solvents. |
IUPAC Name | 2-diethylphosphanylethyl(diethyl)phosphane |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'