missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,2-Bis(diethylphosphino)ethane, 98%

Brand:  Alfa Aesar™ 039272.MD

Code : NEW

Additional Details : CAS Number : 6411-21-8 Weight : 0.00025kg Transport : UN number : 2845 Chem class : 4.2 Pack group : I

Quantity 250mg

 View more versions of this product

Product Code. 15481527

  • £22.37 / 250mg


Estimated Shipment date
from Supplier 20-07-2018
Add to basket

Description

Description

  • Packaged under argon
Specifications

Chemical Identifiers

1,2-Bis(diethylphosphino)ethane
(CH3CH2)2PCH2CH2P(CH2CH3)2
MFCD00015175
6411-21-8
206.25
UN2845

Specifications

1,2-Bis(diethylphosphino)ethane
Liquid
(CH3CH2)2PCH2CH2P(CH2CH3)2
MFCD00015175
0.884g/mL
124° to 126°C (10mmHg)
Pungent
6411-21-8
98%
206.25
UN2845
Colorless
250mg
Air Sensitive
Videos
SDS