missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 1,2-Bis(carboxymethylthio)ethane, 97%
Click to view available options
Quantity:
5 g
25 g
Specifications
Melting Point | 110°C |
Molecular Formula | C6H8O4S2 |
MDL Number | MFCD00021771 |
Quantity | 5 g |
Synonym | ethylenedithio diacetic acid, 1,2-bis carboxymethylthio ethane, 2-2-carboxymethylsulfanyl ethylsulfanyl acetic acid, 2-2-carboxymethylthio ethylthio acetic acid, 2-carboxymethyl sulfanyl ethyl sulfanyl acetic acid, ethylenedithiodiacetic acid, acmc-2097kv, ethylenebisthio diacetic acid, 1,2-ethylenebis thioglycolic acid, 2,2'-ethylenedithio diacetic acid |
InChI Key | CCQMORVULNZXIA-UHFFFAOYSA-L |
SMILES | C(CSCC(=O)O)SCC(=O)O |
IUPAC Name | 2-[2-(carboxymethylsulfanyl)ethylsulfanyl]acetic acid |
Molecular Weight (g/mol) | 208.25 |
PubChem CID | 310815 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'